The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000724.3 | NP_000715.2 | 12 - 13 | 1936 | 103 |
NM_001167945.1 | NP_001161417.1 | 13 - 14 | 1655 | 103 | |
NM_201570.2 | NP_963864.1 | 12 - 13 | 1790 | 103 | |
NM_201571.3 | NP_963865.2 | 13 - 14 | 1769 | 103 | |
NM_201572.3 | NP_963866.2 | 13 - 14 | 1697 | 103 | |
NM_201590.2 | NP_963884.2 | 12 - 13 | 1566 | 103 | |
NM_201593.2 | NP_963887.2 | 13 - 14 | 1432 | 103 | |
NM_201596.2 | NP_963890.2 | 13 - 14 | 1546 | 103 | |
NM_201597.2 | NP_963891.1 | 13 - 14 | 1474 | 103 | |
XM_005252588.3 | XP_005252645.1 | 12 - 13 | 1680 | 103 | |
XM_005252591.3 | XP_005252648.1 | 7 - 8 | 741 | 103 | |
XM_006717502.3 | XP_006717565.1 | 12 - 13 | 1599 | 103 | |
XM_011519659.2 | XP_011517961.1 | 12 - 13 | 1494 | 103 | |
XM_011519660.2 | XP_011517962.1 | 12 - 13 | 2037 | 103 | |
XM_017016625.1 | XP_016872114.1 | 13 - 14 | 1654 | 103 | |
GenBank mRNA | AB208917.1 | - | 12 - 13 | 1822 | 103 |
AF137376.1 | - | 13 - 14 | 1484 | 103 | |
AF137377.1 | - | 12 - 13 | 1327 | 103 | |
AF285239.2 | - | 12 - 13 | 1324 | 103 | |
AF423189.1 | - | 12 - 13 | 1321 | 103 | |
AF423190.1 | - | 13 - 14 | 1402 | 103 | |
AF423191.1 | - | 13 - 14 | 1486 | 103 | |
AF423192.1 | - | 12 - 13 | 1342 | 103 | |
AF465485.1 | - | 12 - 13 | 1216 | 103 | |
AK293937.1 | - | 12 - 13 | 1237 | 103 | |
AK296471.1 | - | 13 - 14 | 1453 | 103 | |
AK299873.1 | - | 12 - 13 | 1692 | 103 | |
AK307526.1 | - | 13 - 14 | 1655 | 103 | |
AY393858.1 | - | 13 - 14 | 1402 | 103 | |
AY393859.1 | - | 12 - 13 | 1324 | 103 | |
AY393860.1 | - | 13 - 14 | 1414 | 103 | |
AY393861.1 | - | 13 - 14 | 1330 | 103 | |
AY393862.1 | - | 12 - 13 | 1252 | 103 | |
AY675091.1 | - | 6 - 7 | 736 | 103 | |
BC136409.1 | - | 13 - 14 | 1509 | 103 | |
U95019.1 | - | 12 - 13 | 1822 | 103 |