The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024654.4 | NP_078930.3 | 10 - 11 | 1920 | 68 |
XM_005263493.3 | XP_005263550.1 | 8 - 9 | 1252 | 68 | |
XM_011542147.2 | XP_011540449.1 | 5 - 6 | 846 | 68 | |
XM_017002338.1 | XP_016857827.1 | 8 - 9 | 1430 | 68 | |
XM_017002339.1 | XP_016857828.1 | 7 - 8 | 1146 | 68 | |
GenBank mRNA | AK026976.1 | - | 6 - 7 | 930 | 68 |
AK091284.1 | - | 10 - 11 | 1857 | 68 | |
AK289653.1 | - | 10 - 11 | 1852 | 68 | |
BC009257.2 | - | 7 - 8 | 1010 | 68 | |
BC012439.1 | - | 3 - 4 | 496 | 68 | |
BC063639.1 | - | 7 - 8 | 960 | 68 | |
BC073860.1 | - | 6 - 7 | 923 | 68 | |
BC094836.1 | - | 9 - 10 | 1295 | 68 | |
BC105095.1 | - | 10 - 11 | 1920 | 68 | |
BC110849.1 | - | 10 - 11 | 1829 | 68 | |
BC112278.1 | - | 10 - 11 | 1920 | 68 |