The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004582.3 | NP_004573.2 | 2 - 3 | 188 | 154 |
NR_073562.1 | 2 - 3 | 188 | 154 | ||
GenBank mRNA | AB209023.1 | - | 1 - 2 | 110 | 154 |
AK293057.1 | - | 2 - 3 | 188 | 154 | |
BC020790.1 | - | 2 - 3 | 181 | 154 | |
CR456941.1 | - | 1 - 2 | 110 | 154 | |
U49245.1 | - | 1 - 2 | 110 | 154 | |
X98001.1 | - | 1 - 2 | 134 | 154 | |
Y08201.1 | - | 1 - 2 | 113 | 154 |