The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256849.1 | NP_001243778.1 | 3 - 4 | 387 | 139 |
NM_001308632.1 | NP_001295561.1 | 2 - 3 | 315 | 139 | |
NM_002691.3 | NP_002682.2 | 3 - 4 | 384 | 139 | |
NR_046402.1 | 3 - 4 | 384 | 139 | ||
XM_005259008.4 | XP_005259065.1 | 3 - 4 | 416 | 139 | |
XM_011527038.1 | XP_011525340.1 | 3 - 4 | 442 | 139 | |
XM_017026881.1 | XP_016882370.1 | 3 - 4 | 461 | 139 | |
XM_017026882.1 | XP_016882371.1 | 3 - 4 | 377 | 139 | |
GenBank mRNA | AB209560.1 | - | 3 - 4 | 503 | 139 |
BC008800.1 | - | 3 - 4 | 139 | ||
CD101690.1 | - | 3 - 4 | 410 | 139 | |
DQ234348.1 | - | 2 - 3 | 139 | ||
M80397.1 | - | 3 - 4 | 368 | 139 | |
M81735.1 | - | 3 - 4 | 357 | 139 |