The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001729.3 | NP_001720.1 | 4 - 5 | 789 | 153 |
XM_011532211.1 | XP_011530513.1 | 4 - 5 | 770 | 153 | |
GenBank mRNA | BC011618.2 | - | 4 - 5 | 580 | 153 |
KU177946.1 | - | 4 - 5 | 428 | 153 | |
S55606.1 | - | 4 - 5 | 722 | 153 |