The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002804.4 | NP_002795.2 | 2 - 3 | 352 | 85 |
XM_017018026.1 | XP_016873515.1 | 2 - 3 | 352 | 85 | |
GenBank mRNA | AK222485.1 | - | 2 - 3 | 114 | 85 |
AK291896.1 | - | 2 - 3 | 129 | 85 | |
AK313518.1 | - | 2 - 3 | 338 | 85 | |
BC008713.2 | - | 2 - 3 | 316 | 85 | |
BC073165.1 | - | 2 - 3 | 297 | 85 | |
BC106920.2 | - | 2 - 3 | 244 | 85 | |
BC107804.1 | - | 2 - 3 | 316 | 85 | |
BP284685.1 | - | 2 - 3 | 354 | 85 | |
CR456731.1 | - | 1 - 2 | 53 | 85 | |
M34079.1 | - | 2 - 3 | 115 | 85 |