The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032177.3 | NP_115553.2 | 3 - 4 | 878 | 127 |
GenBank mRNA | AK023255.1 | - | 3 - 4 | 878 | 127 |
AK024065.1 | - | 3 - 4 | 846 | 127 | |
AK225464.1 | - | 3 - 4 | 843 | 127 | |
BC021161.2 | - | 3 - 4 | 852 | 127 | |
CR457305.1 | - | 3 - 4 | 830 | 127 |