The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002661.4 | NP_002652.2 | 24 - 25 | 2798 | 106 |
GenBank mRNA | AB208914.1 | - | 24 - 25 | 2754 | 106 |
AK304722.1 | - | 21 - 22 | 2202 | 106 | |
AY364256.1 | - | 23 - 24 | 2584 | 106 | |
BC007565.1 | - | 24 - 25 | 2731 | 106 | |
BC011772.2 | - | 24 - 25 | 2731 | 106 | |
BC014561.2 | - | 24 - 25 | 2731 | 106 | |
BC018646.2 | - | 24 - 25 | 2731 | 106 | |
M37238.1 | - | 24 - 25 | 2736 | 106 | |
X14034.1 | - | 24 - 25 | 2736 | 106 |