The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320272.1 | NP_001307201.1 | 3 - 4 | 567 | 143 |
XM_017000567.1 | XP_016856056.1 | 2 - 3 | 740 | 143 | |
XM_017000568.1 | XP_016856057.1 | 2 - 3 | 261 | 143 | |
XM_017000569.1 | XP_016856058.1 | 2 - 3 | 2265 | 143 | |
GenBank mRNA | AK098092.1 | - | 3 - 4 | 567 | 143 |