Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG6
Entrez Gene ID
59285
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 6
Gene Aliases
-
Chromosome Location
Chr.19: 53991149 - 54012666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145814.1 NP_665813.1 1 - 2 923 143
NM_145815.1 NP_665814.1 1 - 2 923 143
XM_006723316.3 XP_006723379.1 1 - 2 1553 143
GenBank mRNA AF288386.1 - 1 - 2 463 143
AF361352.1 - 1 - 2 333 143
BC021552.1 - 1 - 2 923 143

View Full Product Details