The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318185.1 | NP_001305114.1 | 2 - 3 | 182 | 82 |
NM_005353.2 | NP_005344.2 | 2 - 3 | 182 | 82 | |
XM_011545835.2 | XP_011544137.1 | 2 - 3 | 259 | 82 | |
XM_011545836.2 | XP_011544138.1 | 2 - 3 | 258 | 82 | |
XM_011545837.2 | XP_011544139.1 | 2 - 3 | 261 | 82 | |
XM_011545838.2 | XP_011544140.1 | 2 - 3 | 258 | 82 | |
XM_011545839.2 | XP_011544141.1 | 2 - 3 | 258 | 82 | |
XM_011545841.2 | XP_011544143.1 | 2 - 3 | 257 | 82 | |
XM_011545842.2 | XP_011544144.1 | 2 - 3 | 261 | 82 | |
XM_011545843.2 | XP_011544145.1 | 2 - 3 | 265 | 82 | |
XM_011545844.2 | XP_011544146.1 | 2 - 3 | 260 | 82 | |
XM_011545845.2 | XP_011544147.1 | 2 - 3 | 270 | 82 | |
XM_011545846.2 | XP_011544148.1 | 2 - 3 | 268 | 82 | |
XM_011545847.2 | XP_011544149.1 | 2 - 3 | 258 | 82 | |
XM_011545848.2 | XP_011544150.1 | 2 - 3 | 269 | 82 | |
XM_017023215.1 | XP_016878704.1 | 2 - 3 | 261 | 82 | |
GenBank mRNA | AB208945.1 | - | 2 - 3 | 182 | 82 |
AK097160.1 | - | 2 - 3 | 217 | 82 | |
AK301028.1 | - | 2 - 3 | 159 | 82 | |
U37028.1 | - | 2 - 3 | 133 | 82 |