The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004171.3 | NP_004162.2 | 1 - 2 | 610 | 62 |
XM_017018139.1 | XP_016873628.1 | 1 - 2 | 386 | 62 | |
GenBank mRNA | AY066021.1 | - | 1 - 2 | 301 | 62 |
BC132768.1 | - | 1 - 2 | 177 | 62 | |
D85884.1 | - | 1 - 2 | 95 | 62 | |
Z32517.1 | - | 1 - 2 | 107 | 62 |