The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195728.2 | NP_001182657.1 | 5 - 6 | 789 | 67 |
NM_001252652.1 | NP_001239581.1 | 5 - 6 | 854 | 67 | |
NM_004171.3 | NP_004162.2 | 4 - 5 | 1151 | 67 | |
XM_005253067.1 | XP_005253124.1 | 4 - 5 | 628 | 67 | |
XM_011520285.2 | XP_011518587.1 | 4 - 5 | 665 | 67 | |
XM_017018136.1 | XP_016873625.1 | 4 - 5 | 1448 | 67 | |
XM_017018137.1 | XP_016873626.1 | 4 - 5 | 668 | 67 | |
XM_017018138.1 | XP_016873627.1 | 4 - 5 | 652 | 67 | |
XM_017018139.1 | XP_016873628.1 | 4 - 5 | 927 | 67 | |
GenBank mRNA | AB209444.1 | - | 4 - 5 | 567 | 67 |
AK122843.1 | - | 5 - 6 | 854 | 67 | |
AK298769.1 | - | 4 - 5 | 663 | 67 | |
AY066021.1 | - | 4 - 5 | 842 | 67 | |
BC132768.1 | - | 4 - 5 | 718 | 67 | |
BX509751.1 | - | 4 - 5 | 633 | 67 | |
D85884.1 | - | 4 - 5 | 636 | 67 | |
KP966086.1 | - | 4 - 5 | 559 | 67 | |
U01824.1 | - | 5 - 6 | 710 | 67 | |
U03505.1 | - | 4 - 5 | 592 | 67 | |
Z32517.1 | - | 4 - 5 | 648 | 67 |