The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003585.4 | NP_003576.2 | 1 - 2 | 525 | 68 |
XM_011535363.2 | XP_011533665.1 | 1 - 2 | 416 | 68 | |
XM_017025227.1 | XP_016880716.1 | 1 - 2 | 419 | 68 | |
GenBank mRNA | AI478508.1 | - | 1 - 2 | 42 | 68 |
D70830.1 | - | 1 - 2 | 534 | 68 |