The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016937.3 | NP_058633.2 | 5 - 6 | 457 | 65 |
XM_005274552.3 | XP_005274609.1 | 5 - 6 | 589 | 65 | |
XM_006724499.2 | XP_006724562.1 | 4 - 5 | 399 | 65 | |
XM_011545540.2 | XP_011543842.1 | 5 - 6 | 544 | 65 | |
XM_011545541.2 | XP_011543843.1 | 5 - 6 | 545 | 65 | |
XM_017029594.1 | XP_016885083.1 | 5 - 6 | 545 | 65 | |
XM_017029595.1 | XP_016885084.1 | 5 - 6 | 545 | 65 | |
XM_017029596.1 | XP_016885085.1 | 5 - 6 | 545 | 65 | |
GenBank mRNA | BX648513.1 | - | 5 - 6 | 482 | 65 |
X06745.1 | - | 5 - 6 | 458 | 65 |