The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145263.3 | NP_660306.1 | 4 - 5 | 797 | 75 |
NR_123728.1 | 4 - 5 | 797 | 75 | ||
XM_005265717.2 | XP_005265774.1 | 4 - 5 | 712 | 75 | |
XM_017007731.1 | XP_016863220.1 | 2 - 3 | 195 | 75 | |
GenBank mRNA | AB465501.1 | - | 4 - 5 | 423 | 75 |
AK057468.1 | - | 4 - 5 | 709 | 75 | |
AK314799.1 | - | 4 - 5 | 667 | 75 | |
BC025396.1 | - | 4 - 5 | 704 | 75 |