Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA18
Entrez Gene ID
132671
Gene Name
spermatogenesis associated 18
Gene Aliases
Mieap, SPETEX1
Chromosome Location
Chr.4: 52051331 - 52097305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297608.1 NP_001284537.1 6 - 7 1297 81
NM_145263.3 NP_660306.1 7 - 8 1393 81
NR_123728.1 7 - 8 1393 81
XM_005265716.1 XP_005265773.1 5 - 6 1099 81
XM_005265718.1 XP_005265775.1 3 - 4 881 81
XM_011534369.1 XP_011532671.1 6 - 7 1195 81
XM_011534370.1 XP_011532672.1 7 - 8 1319 81
XM_011534372.1 XP_011532674.1 4 - 5 973 81
XM_017007731.1 XP_016863220.1 5 - 6 791 81
GenBank mRNA AB465501.1 - 7 - 8 1019 81
AB465502.1 - 6 - 7 923 81
AK057468.1 - 7 - 8 1305 81
AK098625.1 - 4 - 5 618 81
AK304386.1 - 6 - 7 1212 81
AK314799.1 - 7 - 8 1263 81
BC025396.1 - 7 - 8 1300 81
BC037886.1 - 1 - 2 72 81

View Full Product Details