The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002103.4 | NP_002094.2 | 3 - 4 | 732 | 59 |
NR_027763.1 | 2 - 3 | 550 | 59 | ||
GenBank mRNA | AK223579.1 | - | 3 - 4 | 688 | 59 |
AK291672.1 | - | 3 - 4 | 688 | 59 | |
AK300369.1 | - | 3 - 4 | 520 | 59 | |
AK302738.1 | - | 2 - 3 | 506 | 59 | |
AK303712.1 | - | 2 - 3 | 506 | 59 | |
BC002617.2 | - | 3 - 4 | 671 | 59 | |
BC007688.2 | - | 3 - 4 | 658 | 59 | |
J04501.1 | - | 3 - 4 | 651 | 59 | |
U32573.1 | - | 3 - 4 | 633 | 59 |