The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138795.3 | NP_620150.1 | 5 - 6 | 612 | 66 |
GenBank mRNA | AB118752.1 | - | 5 - 6 | 441 | 66 |
AK127138.1 | - | 5 - 6 | 565 | 66 | |
AK313122.1 | - | 5 - 6 | 590 | 66 | |
BC015408.1 | - | 5 - 6 | 543 | 66 | |
BM548962.1 | - | 5 - 6 | 597 | 66 |