The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243728.1 | NP_001230657.1 | 7 - 8 | 649 | 121 |
NM_001256418.1 | NP_001243347.1 | 8 - 9 | 740 | 121 | |
NM_022745.4 | NP_073582.3 | 7 - 8 | 788 | 121 | |
GenBank mRNA | AK026004.1 | - | 7 - 8 | 725 | 121 |
AK301874.1 | - | 8 - 9 | 717 | 121 | |
AK302090.1 | - | 7 - 8 | 626 | 121 | |
BC008498.1 | - | 7 - 8 | 719 | 121 | |
BC018781.1 | - | 7 - 8 | 717 | 121 | |
BC071731.1 | - | 7 - 8 | 717 | 121 |