The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001414.3 | NP_001405.1 | 1 - 2 | 232 | 125 |
GenBank mRNA | AK294815.1 | - | 1 - 2 | 224 | 125 |
AL541292.3 | - | 1 - 2 | 200 | 125 | |
BC103763.1 | - | 1 - 2 | 94 | 125 | |
DA221985.1 | - | 1 - 2 | 225 | 125 | |
DA879094.1 | - | 1 - 2 | 232 | 125 |