The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032383.4 | NP_115759.2 | 1 - 2 | 356 | 105 |
XM_005247834.3 | XP_005247891.1 | 1 - 2 | 331 | 105 | |
XM_017007323.1 | XP_016862812.1 | 1 - 2 | 331 | 105 | |
GenBank mRNA | AK291631.1 | - | 1 - 2 | 331 | 105 |
AY033141.1 | - | 1 - 2 | 356 | 105 | |
BC040359.1 | - | 1 - 2 | 293 | 105 |