The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032330.1 | NP_115706.1 | 1 - 1 | 940 | 98 |
GenBank mRNA | AY052551.1 | - | 1 - 1 | 912 | 98 |
BC005397.1 | - | 1 - 1 | 928 | 98 | |
BC006000.1 | - | 1 - 1 | 940 | 98 |