The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018362.3 | NP_060832.1 | 1 - 2 | 66 | 75 |
GenBank mRNA | AK002077.1 | - | 1 - 2 | 60 | 75 |
AK223156.1 | - | 1 - 2 | 60 | 75 | |
BC009774.1 | - | 1 - 2 | 53 | 75 | |
BC053907.1 | - | 1 - 2 | 43 | 75 | |
DA231006.1 | - | 1 - 2 | 66 | 75 |