Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIN7C
Entrez Gene ID
55327
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
Gene Aliases
LIN-7-C, LIN-7C, MALS-3, MALS3, VELI3
Chromosome Location
Chr.11: 27494418 - 27506779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018362.3 NP_060832.1 2 - 3 180 105
GenBank mRNA AK002077.1 - 2 - 3 174 105
AK223156.1 - 2 - 3 174 105
BC009774.1 - 2 - 3 167 105
BC053907.1 - 2 - 3 157 105
DA231006.1 - 2 - 3 180 105

View Full Product Details