The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303276.1 | NP_001290205.1 | 16 - 17 | 1983 | 91 |
NM_024844.4 | NP_079120.1 | 17 - 18 | 2077 | 91 | |
XM_005257692.3 | XP_005257749.1 | 15 - 16 | 1666 | 91 | |
XM_006722094.1 | XP_006722157.1 | 16 - 17 | 1702 | 91 | |
XM_011525267.1 | XP_011523569.1 | 16 - 17 | 1715 | 91 | |
XM_017025108.1 | XP_016880597.1 | 15 - 16 | 1880 | 91 | |
XM_017025109.1 | XP_016880598.1 | 15 - 16 | 1741 | 91 | |
XM_017025110.1 | XP_016880599.1 | 15 - 16 | 1607 | 91 | |
GenBank mRNA | AF498261.1 | - | 17 - 18 | 1765 | 91 |
AF514995.1 | - | 17 - 18 | 1845 | 91 | |
AK022611.1 | - | 17 - 18 | 1807 | 91 | |
AK124501.1 | - | 7 - 8 | 1642 | 91 | |
AK297574.1 | - | 16 - 17 | 1726 | 91 | |
AL833893.1 | - | 17 - 18 | 1770 | 91 | |
BC000697.2 | - | 17 - 18 | 1836 | 91 | |
BM973048.1 | - | 2 - 3 | 91 |