Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP85
Entrez Gene ID
79902
Gene Name
nucleoporin 85
Gene Aliases
FROUNT, Nup75
Chromosome Location
Chr.17: 75205450 - 75235759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024844.4 NP_079120.1 1 - 2 343 126
XM_005257690.2 XP_005257747.1 1 - 2 103 126
XM_006722094.1 XP_006722157.1 1 - 2 103 126
XM_011525267.1 XP_011523569.1 1 - 2 103 126
XM_011525268.1 XP_011523570.1 1 - 2 103 126
XM_017025108.1 XP_016880597.1 1 - 2 338 126
GenBank mRNA AF514995.1 - 1 - 2 111 126
AK022611.1 - 1 - 2 73 126
AK298519.1 - 1 - 2 73 126
AK302426.1 - 1 - 2 103 126
BC000697.2 - 1 - 2 102 126
DC424627.1 - 1 - 2 346 126

View Full Product Details