The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303276.1 | NP_001290205.1 | 8 - 9 | 1080 | 59 |
NM_024844.4 | NP_079120.1 | 9 - 10 | 1174 | 59 | |
XM_005257690.2 | XP_005257747.1 | 9 - 10 | 934 | 59 | |
XM_005257692.3 | XP_005257749.1 | 7 - 8 | 735 | 59 | |
XM_006722094.1 | XP_006722157.1 | 8 - 9 | 799 | 59 | |
XM_011525267.1 | XP_011523569.1 | 8 - 9 | 812 | 59 | |
XM_011525268.1 | XP_011523570.1 | 9 - 10 | 934 | 59 | |
XM_017025108.1 | XP_016880597.1 | 7 - 8 | 977 | 59 | |
XM_017025109.1 | XP_016880598.1 | 7 - 8 | 838 | 59 | |
XM_017025110.1 | XP_016880599.1 | 7 - 8 | 704 | 59 | |
GenBank mRNA | AF498261.1 | - | 9 - 10 | 862 | 59 |
AF514995.1 | - | 9 - 10 | 942 | 59 | |
AK022611.1 | - | 9 - 10 | 904 | 59 | |
AK295694.1 | - | 8 - 9 | 821 | 59 | |
AK297574.1 | - | 8 - 9 | 823 | 59 | |
AK298519.1 | - | 8 - 9 | 782 | 59 | |
AK302426.1 | - | 9 - 10 | 934 | 59 | |
AL833893.1 | - | 9 - 10 | 867 | 59 | |
BC000697.2 | - | 9 - 10 | 933 | 59 | |
DA375719.1 | - | 2 - 3 | 279 | 59 |