The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003042.3 | NP_003033.3 | 12 - 13 | 1730 | 81 |
XM_005265410.4 | XP_005265467.1 | 12 - 13 | 3087 | 81 | |
XM_005265411.4 | XP_005265468.1 | 12 - 13 | 3026 | 81 | |
XM_006713306.3 | XP_006713369.1 | 12 - 13 | 1685 | 81 | |
XM_011534025.2 | XP_011532327.1 | 12 - 13 | 3777 | 81 | |
XM_011534027.2 | XP_011532329.1 | 12 - 13 | 3716 | 81 | |
XM_017007071.1 | XP_016862560.1 | 13 - 14 | 3509 | 81 | |
XM_017007072.1 | XP_016862561.1 | 13 - 14 | 2819 | 81 | |
GenBank mRNA | AK295702.1 | - | 9 - 10 | 1093 | 81 |
BC033904.1 | - | 12 - 13 | 1698 | 81 | |
BC057291.1 | - | 12 - 13 | 1710 | 81 | |
X54673.1 | - | 11 - 12 | 1553 | 81 |