The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003042.3 | NP_003033.3 | 2 - 3 | 261 | 72 |
XM_005265410.4 | XP_005265467.1 | 2 - 3 | 1618 | 72 | |
XM_011534025.2 | XP_011532327.1 | 2 - 3 | 2308 | 72 | |
XM_017007071.1 | XP_016862560.1 | 3 - 4 | 2040 | 72 | |
XM_017007072.1 | XP_016862561.1 | 3 - 4 | 1350 | 72 | |
GenBank mRNA | BC033904.1 | - | 2 - 3 | 229 | 72 |
BC057291.1 | - | 2 - 3 | 241 | 72 | |
X54673.1 | - | 1 - 2 | 84 | 72 |