The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003042.3 | NP_003033.3 | 7 - 8 | 1118 | 109 |
XM_005265410.4 | XP_005265467.1 | 7 - 8 | 2475 | 109 | |
XM_005265411.4 | XP_005265468.1 | 7 - 8 | 2414 | 109 | |
XM_006713306.3 | XP_006713369.1 | 7 - 8 | 1073 | 109 | |
XM_011534025.2 | XP_011532327.1 | 7 - 8 | 3165 | 109 | |
XM_011534027.2 | XP_011532329.1 | 7 - 8 | 3104 | 109 | |
XM_017007071.1 | XP_016862560.1 | 8 - 9 | 2897 | 109 | |
XM_017007072.1 | XP_016862561.1 | 8 - 9 | 2207 | 109 | |
GenBank mRNA | AK295702.1 | - | 4 - 5 | 481 | 109 |
BC033904.1 | - | 7 - 8 | 1086 | 109 | |
BC057291.1 | - | 7 - 8 | 1098 | 109 | |
X54673.1 | - | 6 - 7 | 941 | 109 |