Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
Chromosome Location
Chr.1: 240013776 - 240475189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305424.1 NP_001292353.1 16 - 17 5154 94
NM_020066.4 NP_064450.3 15 - 16 5142 94
XM_017001839.1 XP_016857328.1 16 - 17 3994 94
XM_017001840.1 XP_016857329.1 15 - 16 3394 94
XM_017001841.1 XP_016857330.1 15 - 16 3852 94
XM_017001842.1 XP_016857331.1 14 - 15 3073 94
XM_017001843.1 XP_016857332.1 13 - 14 3061 94
GenBank mRNA AB209153.1 - 16 - 17 3747 94
AF218942.1 - 7 - 8 642 94
AF225426.1 - 7 - 8 664 94
AK297716.1 - 10 - 11 912 94
AK298141.1 - 8 - 9 977 94
BC014364.2 - 5 - 6 481 94

View Full Product Details