The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020066.4 | NP_064450.3 | 3 - 4 | 2150 | 107 |
XM_017001843.1 | XP_016857332.1 | 3 - 4 | 2148 | 107 | |
GenBank mRNA | AF218941.1 | - | 3 - 4 | 740 | 107 |