Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
Chromosome Location
Chr.1: 240013776 - 240475189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020066.4 NP_064450.3 4 - 5 2211 106
GenBank mRNA AF218941.1 - 4 - 5 801 106

View Full Product Details