Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
Chromosome Location
Chr.1: 240013776 - 240475189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305424.1 NP_001292353.1 6 - 7 4163 98
NM_020066.4 NP_064450.3 5 - 6 4151 98
XM_011544237.2 XP_011542539.1 6 - 7 4161 98
XM_017001837.1 XP_016857326.1 6 - 7 4161 98
XM_017001838.1 XP_016857327.1 6 - 7 4161 98
XM_017001839.1 XP_016857328.1 6 - 7 3003 98
XM_017001840.1 XP_016857329.1 5 - 6 2403 98
XM_017001841.1 XP_016857330.1 5 - 6 2861 98
GenBank mRNA AB209153.1 - 6 - 7 2756 98
DN990332.1 - 1 - 2 155 98

View Full Product Details