Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
Chromosome Location
Chr.1: 240013776 - 240475189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305424.1 NP_001292353.1 10 - 11 4540 95
NM_020066.4 NP_064450.3 9 - 10 4528 95
XM_017001839.1 XP_016857328.1 10 - 11 3380 95
XM_017001840.1 XP_016857329.1 9 - 10 2780 95
XM_017001841.1 XP_016857330.1 9 - 10 3238 95
XM_017001842.1 XP_016857331.1 8 - 9 2459 95
XM_017001843.1 XP_016857332.1 7 - 8 2447 95
GenBank mRNA AB209153.1 - 10 - 11 3133 95
AK297716.1 - 4 - 5 298 95
AK297755.1 - 4 - 5 544 95
AK298141.1 - 2 - 3 363 95
DN990332.1 - 5 - 6 532 95

View Full Product Details