Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMC3
Entrez Gene ID
10319
Gene Name
laminin subunit gamma 3
Gene Aliases
OCCM
Chromosome Location
Chr.9: 131009117 - 131093059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006059.3 NP_006050.3 24 - 25 4121 79
GenBank mRNA AB208887.1 - 23 - 24 3437 79
AF041835.1 - 25 - 26 4156 79

View Full Product Details