Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 47850690 - 48089279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134364.1 NP_001127836.1 10 - 11 2845 67
NM_002375.4 NP_002366.2 10 - 11 2845 67
XM_005265133.4 XP_005265190.1 13 - 14 5911 67
XM_005265134.4 XP_005265191.1 13 - 14 6324 67
XM_005265155.4 XP_005265212.1 9 - 10 2769 67
XM_005265157.4 XP_005265214.1 3 - 4 2138 67
XM_005265158.2 XP_005265215.1 3 - 4 2138 67
XM_006713146.3 XP_006713209.1 14 - 15 5943 67
XM_006713147.3 XP_006713210.1 14 - 15 6360 67
XM_006713152.3 XP_006713215.1 13 - 14 6309 67
XM_011533703.2 XP_011532005.1 13 - 14 5860 67
XM_011533704.2 XP_011532006.1 13 - 14 5860 67
XM_011533705.1 XP_011532007.1 12 - 13 5809 67
XM_011533708.2 XP_011532010.1 12 - 13 5809 67
XM_011533709.2 XP_011532011.1 13 - 14 5860 67
XM_011533710.2 XP_011532012.1 12 - 13 5737 67
XM_011533718.1 XP_011532020.1 12 - 13 5809 67
XM_011533719.2 XP_011532021.1 11 - 12 2425 67
XM_017006391.1 XP_016861880.1 13 - 14 5860 67
XM_017006392.1 XP_016861881.1 13 - 14 5860 67
XM_017006393.1 XP_016861882.1 13 - 14 5892 67
XM_017006394.1 XP_016861883.1 12 - 13 6186 67
XM_017006395.1 XP_016861884.1 11 - 12 2425 67
XM_017006396.1 XP_016861885.1 11 - 12 2425 67
XM_017006397.1 XP_016861886.1 11 - 12 2425 67
XM_017006398.1 XP_016861887.1 10 - 11 2374 67
XM_017006399.1 XP_016861888.1 11 - 12 2889 67
XM_017006400.1 XP_016861889.1 11 - 12 2889 67
XM_017006401.1 XP_016861890.1 9 - 10 2266 67
XM_017006402.1 XP_016861891.1 10 - 11 2838 67
XM_017006403.1 XP_016861892.1 11 - 12 2425 67
XM_017006404.1 XP_016861893.1 11 - 12 2425 67
XM_017006405.1 XP_016861894.1 10 - 11 2838 67
XM_017006406.1 XP_016861895.1 11 - 12 2889 67
XM_017006407.1 XP_016861896.1 11 - 12 2425 67
XM_017006408.1 XP_016861897.1 11 - 12 2425 67
XM_017006409.1 XP_016861898.1 10 - 11 2838 67
XM_017006410.1 XP_016861899.1 10 - 11 2766 67
XM_017006411.1 XP_016861900.1 9 - 10 2715 67
XM_017006412.1 XP_016861901.1 9 - 10 2715 67
XM_017006413.1 XP_016861902.1 10 - 11 2838 67
XM_017006414.1 XP_016861903.1 10 - 11 2838 67
XM_017006415.1 XP_016861904.1 10 - 11 2838 67
XM_017006416.1 XP_016861905.1 8 - 9 2215 67
XM_017006417.1 XP_016861906.1 3 - 4 2138 67
XM_017006418.1 XP_016861907.1 3 - 4 2138 67
XM_017006419.1 XP_016861908.1 3 - 4 2138 67
XM_017006420.1 XP_016861909.1 3 - 4 2138 67
XM_017006421.1 XP_016861910.1 3 - 4 2138 67
XM_017006422.1 XP_016861911.1 3 - 4 2138 67
GenBank mRNA AB209377.1 - 10 - 11 2405 67
AK125245.1 - 3 - 4 1671 67
BC008715.2 - 8 - 9 1890 67
BC012794.2 - 8 - 9 1890 67
BC015149.2 - 8 - 9 1890 67
BC048200.1 - 3 - 4 1436 67
BG760540.1 - 4 - 5 541 67
CR749544.1 - 3 - 4 550 67
M64571.1 - 10 - 11 2409 67
U19727.1 - 10 - 11 2394 67

View Full Product Details