Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 47850690 - 48089279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011533703.2 XP_011532005.1 21 - 22 6990 65
XM_011533718.1 XP_011532020.1 18 - 19 6732 65
XM_017006391.1 XP_016861880.1 21 - 22 6945 65
XM_017006392.1 XP_016861881.1 20 - 21 6876 65
XM_017006395.1 XP_016861884.1 19 - 20 3555 65
XM_017006397.1 XP_016861886.1 19 - 20 3510 65
XM_017006400.1 XP_016861889.1 19 - 20 4019 65
XM_017006402.1 XP_016861891.1 18 - 19 3968 65
XM_017006404.1 XP_016861893.1 17 - 18 3348 65
XM_017006405.1 XP_016861894.1 18 - 19 3923 65
XM_017006408.1 XP_016861897.1 17 - 18 3303 65
XM_017006412.1 XP_016861901.1 17 - 18 3845 65
XM_017006414.1 XP_016861903.1 16 - 17 3761 65
XM_017006417.1 XP_016861906.1 11 - 12 3268 65
XM_017006418.1 XP_016861907.1 11 - 12 3223 65
XM_017006420.1 XP_016861909.1 10 - 11 3154 65
XM_017006421.1 XP_016861910.1 9 - 10 3061 65
XM_017006422.1 XP_016861911.1 9 - 10 3016 65
GenBank mRNA CR749544.1 - 9 - 10 1473 65

View Full Product Details