The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019112.3 | NP_061985.2 | 2 - 3 | 293 | 73 |
XM_006722616.1 | XP_006722679.1 | 2 - 3 | 280 | 73 | |
XM_006722617.2 | XP_006722680.1 | 2 - 3 | 280 | 73 | |
XM_011527628.2 | XP_011525930.1 | 2 - 3 | 154 | 73 | |
XM_011527629.1 | XP_011525931.1 | 2 - 3 | 280 | 73 | |
XM_011527630.1 | XP_011525932.1 | 2 - 3 | 280 | 73 | |
XM_011527631.1 | XP_011525933.1 | 2 - 3 | 280 | 73 | |
XM_011527633.2 | XP_011525935.1 | 2 - 3 | 280 | 73 | |
XM_011527634.1 | XP_011525936.1 | 2 - 3 | 280 | 73 | |
XM_011527635.1 | XP_011525937.1 | 2 - 3 | 280 | 73 | |
XM_017026142.1 | XP_016881631.1 | 2 - 3 | 280 | 73 | |
XM_017026143.1 | XP_016881632.1 | 2 - 3 | 280 | 73 | |
GenBank mRNA | AF250238.1 | - | 1 - 2 | 62 | 73 |
AF328787.1 | - | 1 - 2 | 178 | 73 | |
AK301108.1 | - | 2 - 3 | 273 | 73 | |
AK310127.1 | - | 2 - 3 | 280 | 73 | |
AK310909.1 | - | 2 - 3 | 269 | 73 | |
DC405492.1 | - | 2 - 3 | 293 | 73 |