The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198265.1 | NP_937998.1 | 1 - 2 | 221 | 150 |
XM_011528756.2 | XP_011527058.1 | 1 - 2 | 239 | 150 | |
GenBank mRNA | BC033591.1 | - | 1 - 2 | 183 | 150 |