The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012444.2 | NP_036576.1 | 6 - 7 | 686 | 136 |
NM_198265.1 | NP_937998.1 | 5 - 6 | 572 | 136 | |
XM_005260379.3 | XP_005260436.1 | 6 - 7 | 704 | 136 | |
XM_005260380.3 | XP_005260437.1 | 6 - 7 | 704 | 130 | |
XM_005260381.3 | XP_005260438.1 | 6 - 7 | 704 | 130 | |
XM_005260382.4 | XP_005260439.1 | 6 - 7 | 704 | 136 | |
XM_011528756.2 | XP_011527058.1 | 5 - 6 | 590 | 136 | |
XM_011528757.2 | XP_011527059.1 | 6 - 7 | 538 | 136 | |
GenBank mRNA | AF149310.1 | - | 6 - 7 | 626 | 136 |
AF169385.1 | - | 6 - 7 | 701 | 136 | |
BC033591.1 | - | 5 - 6 | 534 | 136 |