Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FANCI
Entrez Gene ID
55215
Gene Name
Fanconi anemia complementation group I
Gene Aliases
KIAA1794
Chromosome Location
Chr.15: 89243954 - 89317132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113378.1 NP_001106849.1 27 - 28 3103 96
NM_018193.2 NP_060663.2 26 - 27 2923 96
XM_011521756.2 XP_011520058.1 27 - 28 3127 96
XM_011521757.2 XP_011520059.1 27 - 28 3123 96
XM_011521764.2 XP_011520066.1 26 - 27 2947 96
XM_011521767.2 XP_011520069.1 27 - 28 3097 96
GenBank mRNA AB058697.1 - 13 - 14 3027 96
AK001581.1 - 20 - 21 2312 96
AK027564.1 - 15 - 16 1810 96
AK055176.1 - 26 - 27 2882 96
BC021859.1 - 4 - 5 445 96
BC140769.1 - 27 - 28 3060 96
BC144483.1 - 27 - 28 3060 96
EF469766.1 - 26 - 27 3013 96
EF567077.1 - 26 - 27 3013 96

View Full Product Details