Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FANCI
Entrez Gene ID
55215
Gene Name
Fanconi anemia complementation group I
Gene Aliases
KIAA1794
Chromosome Location
Chr.15: 89243954 - 89317132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113378.1 NP_001106849.1 31 - 32 3440 67
NM_018193.2 NP_060663.2 30 - 31 3260 67
XM_011521756.2 XP_011520058.1 31 - 32 3464 67
XM_011521757.2 XP_011520059.1 31 - 32 3460 67
XM_011521764.2 XP_011520066.1 30 - 31 3284 67
XM_011521767.2 XP_011520069.1 31 - 32 3434 67
GenBank mRNA AB058697.1 - 17 - 18 3364 67
AK027564.1 - 19 - 20 2147 67
BC021859.1 - 8 - 9 782 67
BC140769.1 - 31 - 32 3397 67
BC144483.1 - 31 - 32 3397 67
EF469766.1 - 30 - 31 3350 67
EF567077.1 - 30 - 31 3350 67

View Full Product Details