Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FANCI
Entrez Gene ID
55215
Gene Name
Fanconi anemia complementation group I
Gene Aliases
KIAA1794
Chromosome Location
Chr.15: 89243954 - 89317132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113378.1 NP_001106849.1 9 - 10 839 68
NM_018193.2 NP_060663.2 9 - 10 839 68
XM_011521756.2 XP_011520058.1 9 - 10 863 68
XM_011521757.2 XP_011520059.1 9 - 10 859 68
XM_011521764.2 XP_011520066.1 9 - 10 863 68
XM_011521767.2 XP_011520069.1 9 - 10 833 68
GenBank mRNA AK001581.1 - 3 - 4 228 68
AK055176.1 - 9 - 10 798 68
BC140769.1 - 9 - 10 796 68
BC144483.1 - 9 - 10 796 68
BM461935.1 - 9 - 10 800 68
EF469766.1 - 8 - 9 749 68
EF567077.1 - 8 - 9 749 68

View Full Product Details