The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012964.2 | NP_001012982.1 | 1 - 2 | 433 | 106 |
NM_001319949.1 | NP_001306878.1 | 1 - 2 | 433 | 106 | |
GenBank mRNA | AY318867.1 | - | 1 - 2 | 242 | 106 |
AY318868.1 | - | 1 - 2 | 228 | 106 | |
AY318869.1 | - | 1 - 2 | 228 | 106 | |
BC015525.2 | - | 1 - 2 | 209 | 106 | |
BM763657.1 | - | 1 - 2 | 252 | 106 | |
BP253007.1 | - | 1 - 2 | 433 | 106 |