Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT5C
Entrez Gene ID
30833
Gene Name
5', 3'-nucleotidase, cytosolic
Gene Aliases
DNT, DNT1, HEL74, P5N2, PN-I, PN-II, UMPH2, cdN, dNT-1
Chromosome Location
Chr.17: 75130225 - 75131965 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045513.1 3 - 4 486 135
GenBank mRNA BC008183.1 - 3 - 4 434 135

View Full Product Details