Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC31
Entrez Gene ID
64427
Gene Name
tetratricopeptide repeat domain 31
Gene Aliases
-
Chromosome Location
Chr.2: 74483032 - 74494564 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022492.4 NP_071937.4 4 - 5 487 119
NR_027749.1 3 - 4 384 119
XM_005264492.1 XP_005264549.1 4 - 5 476 119
XM_011533035.1 XP_011531337.1 4 - 5 476 119
XM_011533036.1 XP_011531338.1 4 - 5 476 119
XM_011533037.1 XP_011531339.1 4 - 5 476 119
XM_011533040.1 XP_011531342.1 3 - 4 425 119
XM_017004706.1 XP_016860195.1 4 - 5 476 119
XM_017004707.1 XP_016860196.1 4 - 5 476 119
XM_017004708.1 XP_016860197.1 3 - 4 381 119
XM_017004709.1 XP_016860198.1 3 - 4 611 119
GenBank mRNA AK022850.1 - 4 - 5 473 119
AK090520.1 - 3 - 4 526 119
AK223355.1 - 4 - 5 471 119
AK225626.1 - 4 - 5 473 119
AK303105.1 - 3 - 4 370 119
BC036017.1 - 4 - 5 471 119
BC045536.1 - 3 - 4 375 119
BC096709.1 - 4 - 5 473 119
BC098365.1 - 4 - 5 473 119

View Full Product Details