The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022492.4 | NP_071937.4 | 6 - 7 | 623 | 96 |
NR_027749.1 | 5 - 6 | 520 | 96 | ||
XM_005264492.1 | XP_005264549.1 | 6 - 7 | 612 | 96 | |
XM_011533035.1 | XP_011531337.1 | 6 - 7 | 612 | 96 | |
XM_011533036.1 | XP_011531338.1 | 6 - 7 | 612 | 96 | |
XM_011533037.1 | XP_011531339.1 | 6 - 7 | 612 | 96 | |
XM_011533040.1 | XP_011531342.1 | 5 - 6 | 561 | 96 | |
XM_017004706.1 | XP_016860195.1 | 6 - 7 | 612 | 96 | |
XM_017004707.1 | XP_016860196.1 | 6 - 7 | 612 | 96 | |
XM_017004708.1 | XP_016860197.1 | 5 - 6 | 517 | 96 | |
XM_017004709.1 | XP_016860198.1 | 5 - 6 | 747 | 96 | |
GenBank mRNA | AK022850.1 | - | 6 - 7 | 609 | 96 |
AK090520.1 | - | 5 - 6 | 662 | 96 | |
AK223355.1 | - | 6 - 7 | 607 | 96 | |
AK225626.1 | - | 6 - 7 | 609 | 96 | |
AK303105.1 | - | 5 - 6 | 506 | 96 | |
BC036017.1 | - | 6 - 7 | 607 | 96 | |
BC045536.1 | - | 5 - 6 | 511 | 96 | |
BC096709.1 | - | 6 - 7 | 609 | 96 | |
BC098365.1 | - | 6 - 7 | 609 | 96 | |
BC099729.1 | - | 4 - 5 | 894 | 96 | |
BX648131.1 | - | 3 - 4 | 1075 | 96 |