The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286423.1 | NP_001273352.1 | 15 - 16 | 1746 | 62 |
NM_001286427.1 | NP_001273356.1 | 12 - 13 | 1263 | 62 | |
NM_024506.4 | NP_078782.3 | 15 - 16 | 1734 | 62 | |
XM_011511813.2 | XP_011510115.1 | 13 - 14 | 1323 | 62 | |
XM_017004895.1 | XP_016860384.1 | 13 - 14 | 1430 | 62 | |
GenBank mRNA | AK055098.1 | - | 10 - 11 | 921 | 62 |
AK074393.1 | - | 9 - 10 | 914 | 62 | |
AK299404.1 | - | 14 - 15 | 1495 | 62 | |
AK300215.1 | - | 12 - 13 | 1246 | 62 | |
AY358639.1 | - | 15 - 16 | 1714 | 62 |